Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coagulation Factor XIV/Protein C Mouse, Alexa Fluor 405, Clone: PC 7, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105113AF405
Description
Coagulation Factor XIV/Protein C Monoclonal specifically detects Coagulation Factor XIV/Protein C in Human samples. It is validated for ELISA.Specifications
Coagulation Factor XIV/Protein C | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PROC | |
Recombinant form of human activated protein C (Drotrecogin alfa, Xigris) | |
0.1 mL | |
Apoptosis, Cancer | |
5624 | |
Human, Nonspecies specific | |
Purified |
ELISA | |
PC 7 | |
ELISA | |
Anticoagulant protein C, APC, Autoprothrombin IIA, Blood coagulation factor XIV, EC 3.4.21, EC 3.4.21.69, PC, PROC1, protein C (inactivator of coagulation factors Va and VIIIa), vitamin K-dependent protein C | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Has specificity for a conformation dependent neoepitope that is expressed in activated protein C upon complex-formation with alpha1-antitrypsin. No reaction is seen to non-complexed alpha1-antitrypsin and only very little cross reaction to protein C zymogen. Note that specificity is calcium dependent. | |
Store at 4C in the dark. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title