Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coagulation Factor XIV/Protein C Mouse anti-Human, Alexa Fluor™ 750, Clone: 3C7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223530AF750

Catalog No. NP22330A70


Add to cart

Description

Description

Coagulation Factor XIV/Protein C Monoclonal antibody specifically detects Coagulation Factor XIV/Protein C in Human samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

Coagulation Factor XIV/Protein C
3C7
Western Blot, ELISA
Anticoagulant protein C, APC, Autoprothrombin IIA, Blood coagulation factor XIV, EC 3.4.21, EC 3.4.21.69, PC, PROC1, protein C (inactivator of coagulation factors Va and VIIIa), vitamin K-dependent protein C
Mouse
IgG2b Kappa
Store at 4°C in the dark.
Monoclonal
Clone 3C7 reacts with human protein C
ELISA, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
PROC
Protein C isolated from human plasma and adsorbed onto aluminum hydroxide gel.
0.1 mL
Primary
5624
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit