Learn More
CoagulationFactorIII/TissueFactor Mouse anti-Human, Janelia Fluor 646, Clone: CLB/TF-1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB10064304JF646
Catalog No. NB064304646
Add to cart
Description
Description
CoagulationFactorIII/TissueFactor Monoclonal antibody specifically detects CoagulationFactorIII/TissueFactor in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CoagulationFactorIII/TissueFactor
CLB/TF-1
Flow Cytometry
Purified
F3
Made to Human CD142
Protein A purified
Cancer
Primary
2152
Human
Flow Cytometry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), FLJ17960, TF, TFA, Thromboplastin, tissue factor
Mouse
IgG1
0.1mL
Store at 4°C in the dark.
Monoclonal
NB100-64304 recognizes human CD142, a 45kD cell surface glycoprotein which is otherwise known as Tissue Factor (TF). CD142 expression can be induced on monocytes, macrophages and endothelial cells by various stimuli including interleukin 1, tumour necrosis factor and endotoxin. CD142 initiates the blood clotting cascade by binding coagulation factor VIIa, which activates factor IX or factor X by specific limited proteolysis. CD142 also plays an important role in inflammation, angiogenesis, and the pathophysiology of atherosclerosis and cancer.