Alfa Aesar™ Coal tar solution, contains 81.0-86.0% alcohol

Manufacturer: Alfa Aesar™ 43671AE

Quantity 100mL

 View more versions of this product

Catalog No. AA43671AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 8007-45-2
    MDL Number MFCD00146482
    UN Number UN1993
    Chemical Name or Material Coal tar solution

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P280-P281-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    CAS 8007-45-2
    TSCA TSCA
    MDL Number MFCD00146482
    Flash Point 13°C (55°F)
    UN Number UN1993
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H350
    Highly flammable liquid and vapor.
    May cause cancer.
    Concentration Contains 81.0 to 86.0% alcohol
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Color Dark brown
    Quantity 100mL
    Odor Alcohol-like
    Chemical Name or Material Coal tar solution

    GHS H Statement
    H225-H350
    Highly flammable liquid and vapor.
    May cause cancer.

    GHS P Statement
    P210-P280-P281-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit