Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coal Tar Topical Solution, USP, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
55 gal.
Packaging:
Amber Glass Bottle
Drum
Poly Drum
65996-92-1,64-17-5
65996-92-1,64-17-5

CO100, 65996-92-1

Spectrum™ Coal Tar Topical Solution, USP is used on the skin to treat the itching, scaling, and flaking. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Quantity 55 gal.
Packaging Drum
CAS Max % 20%,80%
UN Number UN1170