Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coal Tar Topical Solution, USP, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
20 L
4 x 4 L
Packaging:
Amber Glass Bottle
Poly Drum
65996-92-1, 64-17-5
65996-92-1, 64-17-5

CO100, 65996-92-1

Spectrum™ Coal Tar Topical Solution, USP is used on the skin to treat the itching, scaling, and flaking. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 20%, 80%
UN Number UN1170
Grade USP
Packaging Amber Glass Bottle
Quantity 4 L