Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coal Tar, USP, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
5 gal.
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
8007-45-2
8007-45-2

C1290, 8007-45-2

Spectrum™ Coal Tar, USP is used on the skin to treat the itching, scaling, and flaking. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Quantity 5 gal.
Ignition Residue 2%
Packaging Steel Can
CAS Max % 100%