Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coal Tar, USP, Spectrum™ Chemical
SDP

Catalog No. 18601779
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

C1290, 8007-45-2

Spectrum™ Chemical Coal Tar, USP is used on the skin to treat the itching, scaling, and flaking. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8007-45-2

Specifications

CAS Max % 100%
Ignition Residue 2%
Packaging Amber Glass Bottle
Quantity 500 mL
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.