Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coatomer Subunit Delta Mouse anti-Human, Mouse, Rat, Biotin, Clone: 1318, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243608B

Catalog No. NBP243608B


Add to cart

Description

Description

Coatomer Subunit Delta Monoclonal antibody specifically detects Coatomer Subunit Delta in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

Coatomer Subunit Delta
1318
Western Blot
Purified
ARCN1
Recombinant protein encompassing a sequence within the center region of human COPD. The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Western Blot
Biotin
PBS with 0.05% Sodium Azide
Archain, archain 1, archain vesicle transport protein 1, coatomer delta subunit, coatomer protein complex, subunit delta, coatomer protein delta-COP, coatomer subunit delta, COPD, Delta-coat protein, delta-COP
Mouse
IgG1
0.1 ml
Primary
372
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit