Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coatomer Subunit Delta Mouse anti-Human, Mouse, Rat, Canine, Monkey, PE/Atto594, Clone: OTI6A6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270203PEATT594

Catalog No. N270203P594


Add to cart

Description

Description

Coatomer Subunit Delta Monoclonal antibody specifically detects Coatomer Subunit Delta in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

Coatomer Subunit Delta
OTI6A6
Flow Cytometry
Affinity Purified, Conjugated
ARCN1
Full length recombinant protein of human Coatomer Subunit Delta (NP_001646) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Monkey, Mouse, Rat
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
Archain, archain 1, archain vesicle transport protein 1, coatomer delta subunit, coatomer protein complex, subunit delta, coatomer protein delta-COP, coatomer subunit delta, COPD, Delta-coat protein, delta-COP
Mouse
IgG1
0.1 mL
Primary
372.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit