Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt Carbonate Basic, Powder, 45.5%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
45 kg
Packaging:
Fiber Drum
Poly Bottle
12602-23-2
12602-23-2
Grade Reagent
CAS Max % 1
Percent Purity 0.455
Iron (Fe) 0.0001
Lead (Pb) 5e-05
Packaging Fiber Drum
Sodium (Na) 0.006
Sulfate 0.0005
Quantity 45 kg

CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Material Safety Data Sheet (MSDS) prior to use.