Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt Carbonate, Powder, 45.5%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
513-79-1
513-79-1
Grade Reagent
CAS Max % 1
Percent Purity 0.455
Chloride 0.0001
Copper (Cu) 5e-05
Iron (Fe) 5e-05
Lead (Pb) 5e-05
Packaging Poly Pail
Sulfate 0.002
Quantity 12 kg

CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.