Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt Chloride, 6-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
2.5kg
Packaging:
Poly Bottle
7791-13-1
CoCl2·6H2O
7791-13-1
CoCl2·6H2O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 237.93
Physical Form Crystals
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.92kg
Quantity 500g
Chemical Name or Material Cobalt Chloride, 6-Hydrate