Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt Chloride ACS AR Crystal, Macron Fine Chemicals™

Click to view available options
Quantity:
500g
Packaging:
POLYSTORMOR
7791-13-1
CoCl2·6H2O
7791-13-1
CoCl2·6H2O
Formula Weight 237.93
Physical Form Crystalline
Grade ACS
Packaging POLYSTORMOR
Quantity 500g
Chemical Name or Material Cobalt Chloride, 6-Hydrate