Learn More
Cobalt Chromium Iron Nickel Molybdenum Manganese wire, 0.25mm (0.01 in.) dia., Thermo Scientific™
Supplier: Thermo Scientific Chemicals 45783G6
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Cobalt Chromium Iron Nickel Molybdenum Manganese wire | |
1450°C to 1460°C | |
MFCD18785637 | |
Insoluble in water. | |
Wire |
0.25mm (0.01 in.) dia., Hard | |
Co:Cr:Fe:Ni:Mo:Mn; 40:20:15:15:7:2 wt% | |
10 m | |
≈8.1g/5m |
Safety and Handling
GHS H Statement
H334-H372-H351-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
H317-H334-H350-H372
TSCA : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.