Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cobalt foil, 0.127mm (0.005 in.) thick, 50x50mm (2x2 in.), 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 39715KJ

Quantity 5 pieces

 View more versions of this product

Catalog No. AA39715KJ


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7440-48-4
MFCD00010935
Cobalt foil, 0.127mm (0.005 in.) thick, 50x50mm (2x2 in.)

Specifications

≈2.83g/50x50mm
7440-48-4
MFCD00010935
Cobalt foil, 0.127mm (0.005 in.) thick, 50x50mm (2x2 in.)
5 pieces
Foil
Odorless
99.9%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit