Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cobalt foil, 0.1mm (0.004 in.) thick, Puratronic™, 99.995% (metals basis)

Click to view available options
Quantity:
25 x 25mm
50 x 50mm
100 x 100mm
7440-48-4
Co
58.933
MFCD00010935
GUTLYIVDDKVIGB-UHFFFAOYSA-N
kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride
104730
CHEBI:27638
cobalt
[Co]
7440-48-4
Co
58.933
MFCD00010935
GUTLYIVDDKVIGB-UHFFFAOYSA-N
kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride
104730
CHEBI:27638
cobalt
[Co]
Quantity 50 x 50mm
Weight 0.56g/25x25mm
Odor Odorless
Physical Form Foil
Percent Purity 99.995%
Grade Puratronic™
Chemical Name or Material Cobalt foil, 0.1mm (0.004 in.) thick
Assay (metals basis)

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c

H317-H334-H350

EINECSNumber : 231-158-0

TSCA : Yes

Recommended Storage : Ambient temperatures