Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt ICP Standard, 1mL = 10mg Co (10,000ppm Co), Ricca Chemical

Click to view available options
Quantity:
50mL
100mL
500mL
Packaging:
Poly natural
7440-48-4,7697-37-2,7440-48-4
Co
58.933
GUTLYIVDDKVIGB-UHFFFAOYSA-N
104730
CHEBI:27638
cobalt
[Co]
7440-48-4,7697-37-2,7440-48-4
Co
58.933
GUTLYIVDDKVIGB-UHFFFAOYSA-N
104730
CHEBI:27638
cobalt
[Co]
CAS Min % 84.0,12.69,0.85
CAS Max % 88.0,13.21,0.88
Physical Form Liquid
Packaging Poly natural
Concentration or Composition Notes 10,000ppm Co
Equivalence 1 mL = 10 mg Co
Color Pink
Quantity 100mL
Chemical Name or Material Cobalt ICP Standard