Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cobalt(II) 2-methoxyethoxide, 5% w/v in 2-methoxyethanol, 99% (m.b.), packaged under Ar in resealable ChemSeal bottles

Manufacturer:  Alfa Aesar™ 42819AE

Quantity 100mL

 View more versions of this product

Catalog No. AA42819AE


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

142600-61-1
Co(OCH2CH2OCH3)2
MFCD00798554
Cobalt (II) 2-methoxyethoxide
C6H14CoO4
209.11
UN3274

Specifications

5% w/v in 2-methoxyethanol
100mL
Liquid
C6H14CoO4
209.11
UN3274
99%, (metals basis)
Air & Moisture Sensitive
142600-61-1
ChemSeal™ bottles
Co(OCH2CH2OCH3)2
MFCD00798554
Cobalt (II) 2-methoxyethoxide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H360-H226-H302-H312-H332
May damage fertility or the unborn child.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYLENE GLYCOL MONOMETHYL ETHER

storageNote1 :

  • Packaged under Argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit