Cobalt(II) acetate, Pale pink to purple powder, Alfa Aesar™

Manufacturer: Alfa Aesar 2313822

Quantity 100g

 View more versions of this product

Catalog No. AA2313822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 71-48-7
    Physical Form Powder
    Assay Percent Range 32% min. as Co
    Molecular Formula C4H6CoO4
    Formula Weight 177.02
    MDL Number MFCD00008689
    Chemical Name or Material Cobalt(II) acetate, Anhydrous, Co 32% min.

    Specifications

    Physical Form Powder
    Formula Weight 177.02
    Quantity 100g
    Color Pale pink to purple
    DOT Information DOT Class: 9, Packaging Group: III
    Health Hazard 2 Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
    EINECS Number 200-755-8
    Anhydrous or Hydrate Anhydrous
    Odor Odorless
    Assay Percent Range 32% min. as Co
    Molecular Formula C4H6CoO4
    MDL Number MFCD00008689
    CAS 71-48-7
    Health Hazard 1 H334-H350-H360-H317-H341-H400-H410; May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. May cause an allergic skin reaction. Suspected of causing genetic defects. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Chemical Name or Material Cobalt(II) acetate, Anhydrous, Co 32% min.

    Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

    H334-H350-H360-H317-H341-H400-H410; May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. May cause an allergic skin reaction. Suspected of causing genetic defects. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit