Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Cobalt(Ii) Acetate Tetrahydrate, Reagent grade, Honeywell™

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Poly Bottle
6147-53-1
C4H14CoO8
249.081
MFCD00149569
ZBYYWKJVSFHYJL-UHFFFAOYSA-L
cobalt ii acetate tetrahydrate,cobalt acetate tetrahydrate,cobaltous acetate tetrahydrate,cobalt diacetate tetrahydrate,bis acetato tetraquacobalt,octan kobaltnaty czech,acetic acid, cobalt 2+ salt, tetrahydrate,octan kobaltnaty,ccris 9441
61372
cobalt(2+);diacetate;tetrahydrate
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2]
6147-53-1
C4H14CoO8
249.081
MFCD00149569
ZBYYWKJVSFHYJL-UHFFFAOYSA-L
cobalt ii acetate tetrahydrate,cobalt acetate tetrahydrate,cobaltous acetate tetrahydrate,cobalt diacetate tetrahydrate,bis acetato tetraquacobalt,octan kobaltnaty czech,acetic acid, cobalt 2+ salt, tetrahydrate,octan kobaltnaty,ccris 9441
61372
cobalt(2+);diacetate;tetrahydrate
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2]

Reagent Grade

  • ≥95% for general laboratory use.
Linear Formula (CH3COO)2Co · 4H2O
UN Number UN3077
Beilstein 3731397
Appearance Crystals
Formula Weight 249.08g/mol
Grade Reagent
Density 1.705g/cu.cm
Packaging Poly Bottle
Melting Point 298°C
Quantity 50g
Chemical Name or Material Cobalt(Ii) Acetate Tetrahydrate
Show More Show Less

P201-P261-P280-P284-P304 + P340-P308 + P313

H302-H315-H317-H334-H341-H350i-H360F-H410

DOTInformation : 9/PGIII

EINECSNumber : 200-755-8

RTECSNumber : AG3325000