Alfa Aesar™ Cobalt(II) bromide, anhydrous, 97%

Manufacturer: Alfa Aesar™ 2314930

Quantity 250g

 View more versions of this product

Catalog No. AA2314930

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7789-43-7
    Formula Weight 218.75
    MDL Number MFCD00016017
    Merck Index 14,2435
    Chemical Name or Material Cobalt(II) bromide, Anhydrous

    Specifications

    Formula Weight 218.75
    Melting Point 678°C (in N2)
    Health Hazard 2 GHS H Statement
    H334-H351-H302-H315-H319-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Suspected of causing cancer.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Merck Index 14,2435
    Color Green
    Solubility Information Soluble in water, methanol, ethanol, acetone, methyl acetate
    RTECS Number GF9595000
    MDL Number MFCD00016017
    CAS 7789-43-7
    Quantity 250g
    Health Hazard 1 Danger
    Density 4.909g/mL
    TSCA TSCA
    Physical Form Crystalline
    Sensitivity Hygroscopic
    Chemical Name or Material Cobalt(II) bromide, Anhydrous
    Assay 97%

    GHS H Statement
    H334-H351-H302-H315-H319-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Suspected of causing cancer.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.

    GHS P Statement
    P284a-P261-P280-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit