Alfa Aesar™ Cobalt(II) chloride, anhydrous, 97%

Manufacturer: Alfa Aesar™ B2203122

Quantity 100g

 View more versions of this product

Catalog No. AAB2203122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7646-79-9
    Molecular Formula Cl2Co
    Linear Formula CoCl2
    Formula Weight 129.84
    MDL Number MFCD00010938
    UN Number UN3077
    Merck Index 14,2437
    Chemical Name or Material Cobalt(II) chloride, anhydrous

    Specifications

    Formula Weight 129.84
    TSCA TSCA
    Odor Odorless
    Molecular Formula Cl2Co
    Linear Formula CoCl2
    MDL Number MFCD00010938
    UN Number UN3077
    Merck Index 14,2437
    Boiling Point 1049°C
    Melting Point 735°C
    Quantity 100g
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P342+P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    RTECS Number GF9800000
    CAS 7646-79-9
    Health Hazard 2 GHS H Statement
    H334-H350-H360-H341-H302-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    Chemical Name or Material Cobalt(II) chloride, anhydrous
    Assay 97%

    Description

    Applications
    Cobalt(II) chloride is used in humidity indicator in weather instruments. In the anhydrous form, it finds use in electroplating of cobalt, in organic chemistry and is a precursor to cobaltocene, (bis(cyclopentadienyl)cobalt(II), which is a good reducing agent. It also serves as a Lewis acid. Cobalt chloride is an indicator for water in desiccants, owing to the reversible hydration/dehydration coupled with distinct color change. Cobalt chloride is useful for producing invisible ink as it turns blue when heated and becomes invisible once it gets cooled. Cobalt(II) chloride catalyzes cross coupling of aryl halides or vinyl halides with aryl grignard reagents in excellent yields.

    Solubility
    Freely soluble in water and methanol. Soluble in alcohol, acetone, ether, glycerol, and pyridine.

    Notes
    Hygroscopic. Incompatible with strong oxidizing agents.

    GHS H Statement
    H334-H350-H360-H341-H302-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P284a-P261-P280-P342+P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit