Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) chloride hexahydrate, pure, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
7791-13-1
Cl2Co·6H2O
237.93
7791-13-1
Cl2Co·6H2O
237.93
Linear Formula CoCl2·6H2O
Fieser 01,155; 06,127; 11,479
Merck Index 15,2421
Solubility Information Solubility in water: 970g/L (20°C).
Formula Weight 237.93
Percent Purity 98.0 to 102.0% (Complexometry)
Grade Pure
Solubility Solubility in water: 970g/L (20°C).
Melting Point 86.0°C
Quantity 1kg
Chemical Name or Material Cobalt(II) chloride hexahydrate
Show More Show Less

GHS H Statement
May damage fertility.
Harmful if inhaled.
Harmful if swallowed.
Suspected of causing genetic defects.
May cause cancer by inhalation.
May cause an allergic skin reaction.
May cause allergy or ast

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN:

GHS Signal Word: Danger