Cobalt(II) chloride, ultra dry, 99.9% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1366603

Packaging Ampouled under Argon
Quantity 1g

 View more versions of this product

Catalog No. AA1366603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7646-79-9
    Molecular Formula Cl2Co
    Linear Formula CoCl2
    Formula Weight 129.84
    MDL Number MFCD00010938
    UN Number UN3077
    Merck Index 14,2437
    Chemical Name or Material Cobalt(II) chloride, ultra dry

    Specifications

    Formula Weight 129.84
    Quantity 1g
    TSCA TSCA
    Sensitivity Hygroscopic
    MDL Number MFCD00010938
    Health Hazard 1 Danger
    Mesh Size -10 Mesh
    Odor Odorless
    Physical Form Beads
    Molecular Formula Cl2Co
    Linear Formula CoCl2
    CAS 7646-79-9
    UN Number UN3077
    Merck Index 14,2437
    Boiling Point 1049°C
    Color Pale Blue
    Melting Point 735°C
    Health Hazard 2 GHS H Statement
    H334-H350-H360-H341-H302-H317;
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P342+P311-P405-P501a;
    Avoid breathing dust/fume/gas/mist/vapours/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Density 3.367
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    RTECS Number GF9800000
    Packaging Ampouled under Argon
    Chemical Name or Material Cobalt(II) chloride, ultra dry
    Assay 99.9%, (metals basis)

    GHS H Statement
    H334-H350-H360-H341-H302-H317;
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P284a-P261-P280-P342+P311-P405-P501a;
    Avoid breathing dust/fume/gas/mist/vapours/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit