Alfa Aesar™ Cobalt(II) fluoride tetrahydrate, Puratronic™, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 1069306

Quantity 5g

 View more versions of this product

Catalog No. AA1069306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13817-37-3
    Formula Weight 168.99 (96.93 Anhydrous)
    MDL Number MFCD00149656
    UN Number UN2923
    Merck Index 14,2440
    Chemical Name or Material Cobalt(II) fluoride tetrahydrate
    Grade Puratronic™

    Specifications

    Formula Weight 168.99 (96.93 Anhydrous)
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00149656
    Melting Point 200°C (dec.)
    Density 2.19g/mL
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
    Odor Odorless
    CAS 13817-37-3
    Color Pink
    TSCA TSCA
    Physical Form Powder
    UN Number UN2923
    Merck Index 14,2440
    Sensitivity Moisture Sensitive
    Quantity 5g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H314-H318-H351-H317
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Suspected of causing cancer.
    May cause an allergic skin reaction.
    Chemical Name or Material Cobalt(II) fluoride tetrahydrate
    Assay 99.99%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H301-H314-H318-H351-H317
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Suspected of causing cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit