Alfa Aesar™ Cobalt(II) hexafluoro-2,4-pentanedionate hydrate, 97%

Manufacturer: Alfa Aesar™ 4159406

Quantity 5g

 View more versions of this product

Catalog No. AA4159406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19648-83-0
    Formula Weight 473.04 (Anhydrous)
    MDL Number MFCD00042510
    Synonym Cobalt(II) hexafluoroacetylacetonate; Bis(1,1,1,5,5,5-hexafluoroacetylacetonato)cobalt(II)
    Chemical Name or Material Cobalt(II) hexafluoro-2,4-pentanedionate hydrate

    Specifications

    Formula Weight 473.04 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P280h-P262-P305+P351+P338
    Wear protective gloves/clothing.
    Do not get in eyes, on skin, or on clothing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Odor Odorless
    Synonym Cobalt(II) hexafluoroacetylacetonate; Bis(1,1,1,5,5,5-hexafluoroacetylacetonato)cobalt(II)
    Melting Point 173° to 179°C
    Physical Form Powder
    MDL Number MFCD00042510
    Color Orange
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H351-H315-H319-H317-H335
    Suspected of causing cancer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    CAS 19648-83-0
    Quantity 5g
    Chemical Name or Material Cobalt(II) hexafluoro-2,4-pentanedionate hydrate
    Assay 97%

    GHS H Statement
    H351-H315-H319-H317-H335
    Suspected of causing cancer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P262-P305+P351+P338
    Wear protective gloves/clothing.
    Do not get in eyes, on skin, or on clothing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit