Alfa Aesar™ Cobalt(II) hydroxide, 97%

Manufacturer: Alfa Aesar™ B2204530

Quantity 250g

 View more versions of this product

Catalog No. AAB2204530

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 21041-93-0
    Molecular Formula H2CoO2
    Linear Formula Co(OH)2
    Formula Weight 92.95
    MDL Number MFCD00016015
    Merck Index 14,2442
    Chemical Name or Material Cobalt(II) hydroxide

    Specifications

    Formula Weight 92.95
    Odor Odorless
    MDL Number MFCD00016015
    Melting Point 250°C (dec.)
    Health Hazard 2 GHS H Statement
    H302-H317
    Harmful if swallowed.
    May cause an allergic skin reaction.
    RTECS Number GG0904500
    TSCA TSCA
    Molecular Formula H2CoO2
    Linear Formula Co(OH)2
    CAS 21041-93-0
    Merck Index 14,2442
    Quantity 250g
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P312-P363-P321-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Cobalt(II) hydroxide
    Assay 97%

    GHS H Statement
    H302-H317
    Harmful if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P301+P312-P363-P321-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit