Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II,III) oxide, 99% (metals basis), Alfa Aesar™

S.A. 40-70m2/g

Manufacturer:  Alfa Aesar 4466022

Quantity 100g

 View more versions of this product

Catalog No. AA4466022

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

1308-06-1
99% (Metal basis)
240.8
Cobalt(II, III) oxide
Powder
Co3O4
MFCD00010939

Specifications

1308-06-1
99% (Metal basis)
240.8
MFCD00010939
Black
Cobalt(II, III) oxide
Powder
Co3O4
Black powder
Odorless
100g

Safety and Handling

H351-H400-H410-H317; Suspected of causing cancer. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. May cause an allergic skin reaction.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 215-157-2

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit