Learn More
Cobalt(II,III) oxide, nanopowder, 99% (metals basis), Thermo Scientific Chemicals
Description
Cobalt(II,III) oxide is used in enamels, pigments, porcelain colorants, semiconductors, ceramics, catalysis, grinding wheels and other fields as an important inorganic material. It is a useful oxidant in chemical industry and as catalyst for organic synthesis. It is widely used as catalyst carrier and as electrode active material. It is also used in senior goggles, other filter materials, in temperature and gas sensors, in electronic ceramics, in lithium ion battery electrode materials, magnetic materials, electrochromic devices and solar energy absorbers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 900°C (decomposition) |
Color | Black |
Physical Form | Powder |
Quantity | 25 g |
UN Number | UN3077 |
Merck Index | 14,2429 |
Solubility Information | Soluble in acids and alkali. Insoluble in water. |
Formula Weight | 240.8 |
Percent Purity | 99% |
Density | 6.11 g/mL |
Show More |
Safety and Handling
Hazard Category | H317-H334-H350i |
Hazard Statement | GHS H Statement H351-H317 Suspected of causing cancer. May cause an allergic skin reaction. |
Precautionary Statement | P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c |
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
EINECSNumber | 215-157-2 |
RTECSNumber | GG2500000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.