Alfa Aesar™ Cobalt(II) iodide, anhydrous, 99.5% (metals basis)

Moisture and humidity indicator, catalyst

Manufacturer: Alfa Aesar™ 2311914

Quantity 25g

 View more versions of this product

Catalog No. AA2311914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15238-00-3
    Formula Weight 312.74
    MDL Number MFCD00016030
    Merck Index 14,2443
    Chemical Name or Material Cobalt(II) iodide, Anhydrous

    Specifications

    Formula Weight 312.74
    Boiling Point 570°C (vac.)
    Melting Point 515°C (vac.)
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Powder
    MDL Number MFCD00016030
    TSCA TSCA
    Sensitivity Light Sensitive & Hygroscopic
    Merck Index 14,2443
    Solubility Information Soluble in water
    Health Hazard 1 Warning
    Mesh Size −60 Mesh
    Odor Odorless
    CAS 15238-00-3
    Quantity 25g
    Health Hazard 2 GHS H Statement
    H341-H317
    Suspected of causing genetic defects.
    May cause an allergic skin reaction.
    Chemical Name or Material Cobalt(II) iodide, Anhydrous
    Assay 99.5%, (metals basis)

    GHS H Statement
    H341-H317
    Suspected of causing genetic defects.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P281-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit