Alfa Aesar™ Cobalt(II) meso-tetrakis(4-methoxyphenyl)porphine

Manufacturer: Alfa Aesar™ H2594506

Quantity 5g

 View more versions of this product

Catalog No. AAH2594506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 28903-71-1
    Molecular Formula C48H36CoN4O4
    Formula Weight 791.79
    MDL Number MFCD00010724
    Synonym 5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine cobalt(II)
    Chemical Name or Material Cobalt(II) meso-tetrakis(4-methoxyphenyl)porphine

    Specifications

    Formula Weight 791.79
    Molecular Formula C48H36CoN4O4
    MDL Number MFCD00010724
    CAS 28903-71-1
    Synonym 5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine cobalt(II)
    Melting Point >300°C
    Quantity 5g
    Chemical Name or Material Cobalt(II) meso-tetrakis(4-methoxyphenyl)porphine

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit