Alfa Aesar™ Cobalt(II) molybdenum oxide, 99.9% (metals basis), Ni 0.5% max

Manufacturer: Alfa Aesar™ 4137618

Quantity 50g

 View more versions of this product

Catalog No. AA4137618

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13762-14-6
    Formula Weight 218.87
    MDL Number MFCD00016019
    UN Number UN3077
    Synonym Cobalt(II) molybdate
    Chemical Name or Material Cobalt(II) molybdenum oxide

    Specifications

    Formula Weight 218.87
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H301-H351-H319-H317-H335
    Toxic if swallowed.
    Suspected of causing cancer.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    RTECS Number QA4730000
    Mesh Size −325 Mesh
    Quantity 50g
    TSCA TSCA
    Physical Form Powder
    UN Number UN3077
    Synonym Cobalt(II) molybdate
    Color Green
    MDL Number MFCD00016019
    CAS 13762-14-6
    Health Hazard 1 Danger
    Concentration Ni 0.5% max.
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Chemical Name or Material Cobalt(II) molybdenum oxide
    Assay 99.9%, (metals basis)

    GHS H Statement
    H301-H351-H319-H317-H335
    Toxic if swallowed.
    Suspected of causing cancer.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit