Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cobalt(II) molybdenum oxide, 99.9% (metals basis), Ni 0.5% max

Manufacturer:  Alfa Aesar™ 4137609

Quantity 10g

 View more versions of this product

Catalog No. AA4137609

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

13762-14-6
MFCD00016019
Cobalt(II) molybdate
218.87
UN3077
Cobalt(II) molybdenum oxide

Specifications

13762-14-6
218.87
UN3077
Ni 0.5% max.
Green
Cobalt(II) molybdenum oxide
Powder
MFCD00016019
Cobalt(II) molybdate
−325 Mesh
10g
99.9%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H351-H319-H317-H335
Toxic if swallowed.
Suspected of causing cancer.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : QA4730000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit