Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cobalt(II) nitrate hexahydrate, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1069403

Quantity 1g

 View more versions of this product

Catalog No. AA1069403

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

10026-22-9
MFCD00149647
14,2444
Puratronic™
291.04 (182.94 Anhydrous)
UN1477
Cobalt(II) nitrate hexahydrate

Specifications

10026-22-9
291.04 (182.94 Anhydrous)
UN1477
1.87g/mL
1.52
55° to 56°C
Cobalt(II) nitrate hexahydrate
Puratronic™
Wet Crystalline Aggregates
MFCD00149647
14,2444
Odorless
Red
1g
99.999%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H272-H334-H350-H360-H341-H302-H317
May intensify fire; oxidizer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P221-P210-P284a-P342+P311-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

RTECSNumber : QU7355500

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit