Alfa Aesar™ Cobalt(II) oxalate, anhydrous, 98%

Manufacturer: Alfa Aesar™ B2295618

Quantity 50g

 View more versions of this product

Catalog No. AAB2295618

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 814-89-1
    Molecular Formula C2CoO4
    Formula Weight 146.95
    MDL Number MFCD00050793
    Merck Index 14,2445
    Chemical Name or Material Cobalt(II) oxalate, anhydrous

    Specifications

    Formula Weight 146.95
    Melting Point >300°C
    Quantity 50g
    TSCA TSCA
    Molecular Formula C2CoO4
    MDL Number MFCD00050793
    CAS 814-89-1
    Merck Index 14,2445
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H332-H317
    Toxic if swallowed.
    Harmful if inhaled.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Cobalt(II) oxalate, anhydrous
    Assay 98%

    GHS H Statement
    H301-H332-H317
    Toxic if swallowed.
    Harmful if inhaled.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P301+P310-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit