Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cobalt(II) oxalate, anhydrous, 98%

Click to view available options
Quantity:
10g
50g
814-89-1
C2CoO4
146.951
MFCD00050793
MULYSYXKGICWJF-UHFFFAOYSA-L
cobalt oxalate,cobalt oxalate 1:1,cobalt oxalate coc2o4,cobalt; oxalic acid,oxalic acid cobalt 2+ salt 1:1,ethanedioic acid, cobalt 2+ salt 1:1,oxalic acid cobalt,cobalt ii oxalate,anhydrous,oxalic acid, cobalt 2+ salt 1:1
69946
cobalt(2+);oxalate
C(=O)(C(=O)[O-])[O-].[Co+2]
814-89-1
C2CoO4
146.951
MFCD00050793
MULYSYXKGICWJF-UHFFFAOYSA-L
cobalt oxalate,cobalt oxalate 1:1,cobalt oxalate coc2o4,cobalt; oxalic acid,oxalic acid cobalt 2+ salt 1:1,ethanedioic acid, cobalt 2+ salt 1:1,oxalic acid cobalt,cobalt ii oxalate,anhydrous,oxalic acid, cobalt 2+ salt 1:1
69946
cobalt(2+);oxalate
C(=O)(C(=O)[O-])[O-].[Co+2]
Merck Index 14,2445
Formula Weight 146.95
Percent Purity 98%
Melting Point >300°C
Quantity 10g
Chemical Name or Material Cobalt(II) oxalate, anhydrous

GHS H Statement
H301-H332-H317
Toxic if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P301+P310-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA