Learn More
Cobalt(II) oxalate dihydrate, Reagent Grade, Thermo Scientific Chemicals
Description
Cobalt(II) oxalate dihydrate is used as a stabilizer for hydrogen cyanide and as a temperature indicator. It is used in the preparation of cobalt catalysts, and cobalt metal powder for powder-metallurgical applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 229°C to 231°C (decomposition) |
Density | 3.021 g/mL |
Odor | Odorless |
Quantity | 100 g |
UN Number | UN3288 |
Merck Index | 14,2445 |
Solubility Information | Soluble in aqueous ammonia; Insoluble in water; Slightly soluble in acids. |
Formula Weight | 182.98 (146.95 Anhydrous) |
Grade | Reagent |
Physical Form | Powder |
Show More |
Safety and Handling
Hazard Category | H302+H312-H350 |
Hazard Statement | GHS H Statement H302-H312 Harmful if swallowed. Harmful in contact with skin. |
Precautionary Statement | P201-P202-P264b-P270-P281-P301+P312-P302+P352-P308+P313-P312-P330-P363-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S. |
EINECSNumber | 212-409-3 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.