Alfa Aesar™ Cobalt(II) oxalate dihydrate, Reagent Grade

Stabilizer for HCN, temperature indicator

Manufacturer: Alfa Aesar™ 8775836

Quantity 500g

 View more versions of this product

Catalog No. AA8775836

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5965-38-8
    Molecular Formula CoC2O4·2H2O
    Formula Weight 182.98 (146.95 Anhydrous)
    MDL Number MFCD00050793
    UN Number UN3288
    Merck Index 14,2445
    Chemical Name or Material Cobalt (II) oxalate dihydrate
    Grade Reagent Grade

    Specifications

    Formula Weight 182.98 (146.95 Anhydrous)
    Health Hazard 3 GHS P Statement
    P280-P301+P312-P312-P363-P322-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Wash contaminated clothing before reuse.
    Specific measures (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Powder
    Health Hazard 2 GHS H Statement
    H302-H312
    Harmful if swallowed.
    Harmful in contact with skin.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    CAS 5965-38-8
    Health Hazard 1 Warning
    Density 3.021g/mL
    Odor Odorless
    Melting Point 229° to 231°C (dec.)
    TSCA TSCA
    Molecular Formula CoC2O4·2H2O
    MDL Number MFCD00050793
    UN Number UN3288
    Merck Index 14,2445
    Solubility Information Soluble in aqueous ammonia; Insoluble in water; Slightly soluble in acids
    Quantity 500g
    Chemical Name or Material Cobalt (II) oxalate dihydrate
    Grade Reagent Grade

    GHS H Statement
    H302-H312
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P280-P301+P312-P312-P363-P322-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Wash contaminated clothing before reuse.
    Specific measures (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit