Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cobalt(II) oxide, 99.998% (metals basis)

Manufacturer:  Alfa Aesar™ 4590706

Quantity 5g

 View more versions of this product

Catalog No. AA4590706

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

1307-96-6
MFCD00016031
14,2446
74.93
UN2811
Cobalt (II) oxide

Specifications

1307-96-6
74.93
UN2811
CoO, may contain up to 10% Co3O4
3 to 7mm
Green-brown
5g
99.998%, (metals basis)
Crystals
MFCD00016031
14,2446
6.45g/mL
Air Sensitive
1935°C
Cobalt (II) oxide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H351-H302-H317
Suspected of causing cancer.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P281-P301+P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : GG2800000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit