Alfa Aesar™ Cobalt(II) perchlorate hexahydrate, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 4432018

Quantity 50g

 View more versions of this product

Catalog No. AA4432018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13478-33-6
    Molecular Formula Co(ClO4)2·6H2O
    Formula Weight 366.74 (257.83 Anhydrous)
    MDL Number MFCD00150207
    UN Number UN1481
    Chemical Name or Material Cobalt (II) perchlorate hexahydrate

    Specifications

    Formula Weight 366.74 (257.83 Anhydrous)
    Health Hazard 2 GHS H Statement
    H272-H351-H315-H319-H317-H335
    May intensify fire
    oxidizer.
    Suspected of causing cancer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 13478-33-6
    Health Hazard 1 Danger
    Odor Odorless
    Sensitivity Hygroscopic
    Molecular Formula Co(ClO4)2·6H2O
    UN Number UN1481
    Quantity 50g
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
    TSCA TSCA
    Physical Form Crystalline
    MDL Number MFCD00150207
    Chemical Name or Material Cobalt (II) perchlorate hexahydrate
    Assay 99.999%, (metals basis)

    GHS H Statement
    H272-H351-H315-H319-H317-H335
    May intensify fire
    oxidizer.
    Suspected of causing cancer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.

    GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit