Alfa Aesar™ Cobalt(II) perchlorate hexahydrate, Reagent Grade

Manufacturer: Alfa Aesar™ 1165209

Quantity 10g

 View more versions of this product

Catalog No. AA1165209

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13478-33-6
    Formula Weight 366.74 (257.83 Anhydrous)
    MDL Number MFCD00150207
    UN Number UN1481
    Chemical Name or Material Cobalt(II) perchlorate hexahydrate
    Grade Reagent Grade

    Specifications

    Formula Weight 366.74 (257.83 Anhydrous)
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H272-H351-H315-H319-H317-H335
    May intensify fire
    oxidizer.
    Suspected of causing cancer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    MDL Number MFCD00150207
    TSCA TSCA
    Odor Odorless
    Physical Form Crystalline
    Sensitivity Hygroscopic
    UN Number UN1481
    Quantity 10g
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
    CAS 13478-33-6
    Chemical Name or Material Cobalt(II) perchlorate hexahydrate
    Grade Reagent Grade

    GHS H Statement
    H272-H351-H315-H319-H317-H335
    May intensify fire
    oxidizer.
    Suspected of causing cancer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.

    GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit