Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cobalt(II) selenide, 99+% (metals basis)

Manufacturer:  Alfa Aesar™ 4022430

Quantity 250g

 View more versions of this product

Catalog No. AA4022430


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1307-99-9
MFCD00016020
Cobalt(II) selenide
137.89
UN3283

Specifications

1307-99-9
137.89
UN3283
7.650g/mL
250g
≥99%, (metals basis)
Powder
MFCD00016020
Soluble in HNO3, aqua regia
Weak
Cobalt(II) selenide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P261-P301+P310-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit