Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cobalt(II) stearate, Co 9-10%

Manufacturer:  Alfa Aesar™ 43352A1

Quantity 1kg

 View more versions of this product

Catalog No. AA43352A1


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1002-88-6
(C18H35O2)2Co
MFCD00054343
C36H70CoO4
625.89
Cobalt (II) stearate

Specifications

Co 9 to 10%
1kg
1.71g/mL
C36H70CoO4
625.89
Cobalt (II) stearate
Purple
1002-88-6
Pellet
(C18H35O2)2Co
MFCD00054343

Safety and Handling

SDS & Safety Documents

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : RG1212000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit