Alfa Aesar™ Cobalt(II) stearate, Co 9-10%

Manufacturer: Alfa Aesar™ 4335230

Quantity 250g

 View more versions of this product

Catalog No. AA4335230

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1002-88-6
    Molecular Formula C36H70CoO4
    Linear Formula (C18H35O2)2Co
    Formula Weight 625.89
    MDL Number MFCD00054343
    Chemical Name or Material Cobalt (II) stearate

    Specifications

    Physical Form Pellet
    Formula Weight 625.89
    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P342+P311-P363-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Wash contaminated clothing before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Linear Formula (C18H35O2)2Co
    Concentration Co 9 to 10%
    Quantity 250g
    Health Hazard 2 GHS H Statement
    H334-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause an allergic skin reaction.
    Density 1.71g/mL
    RTECS Number RG1212000
    TSCA TSCA
    MDL Number MFCD00054343
    Color Purple
    Molecular Formula C36H70CoO4
    CAS 1002-88-6
    Health Hazard 1 Danger
    Chemical Name or Material Cobalt (II) stearate

    GHS H Statement
    H334-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause an allergic skin reaction.

    GHS P Statement
    P284a-P261-P280-P342+P311-P363-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Wash contaminated clothing before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit