Alfa Aesar™ Cobalt(II) sulfate heptahydrate, Puratronic™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 1069614

Quantity 25g

 View more versions of this product

Catalog No. AA1069614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10026-24-1
    Linear Formula CoSO4·7H2O
    Formula Weight 281.10 (154.99 Anhydrous)
    MDL Number MFCD00149658
    UN Number UN3077
    Merck Index 14,2448
    Chemical Name or Material Cobalt(II) sulfate heptahydrate
    Grade Puratronic™

    Specifications

    Formula Weight 281.10 (154.99 Anhydrous)
    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P342+P311-P405-P501a
    Wear respiratory protection.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H334-H350-H360-H341-H302-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    Odor Odorless
    Color Dark red
    MDL Number MFCD00149658
    Quantity 25g
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    UN Number UN3077
    Merck Index 14,2448
    Health Hazard 1 Danger
    Density 2.03g/mL
    Physical Form Crystalline Aggregates
    CAS 10026-24-1
    pH 4
    TSCA TSCA
    Linear Formula CoSO4·7H2O
    RTECS Number GG3200000
    Chemical Name or Material Cobalt(II) sulfate heptahydrate
    Assay 99.999%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H334-H350-H360-H341-H302-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P284a-P261-P280-P342+P311-P405-P501a
    Wear respiratory protection.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit