Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cobalt(II) sulfate heptahydrate, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1069606

Quantity 5g

 View more versions of this product

Catalog No. AA1069606


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

10026-24-1
281.10 (154.99 Anhydrous)
UN3077
Cobalt(II) sulfate heptahydrate
CoSO4·7H2O
MFCD00149658
14,2448
Puratronic™

Specifications

10026-24-1
CoSO4·7H2O
MFCD00149658
14,2448
Odorless
4
Cobalt(II) sulfate heptahydrate
Puratronic™
Crystalline Aggregates
281.10 (154.99 Anhydrous)
UN3077
2.03g/mL
Dark red
5g
99.999%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P342+P311-P405-P501a
Wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : GG3200000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit