Learn More
Cobalt(II) sulfate hydrate, 99.999%, (trace metal basis), ACROS Organics™
Description
Specifications
CAS | 60459-08-7 |
Assay Percent Range | 99.999% |
Molecular Formula | CoH2O5S |
Linear Formula | Co(II)SO4·xH2O |
MDL Number | MFCD00149657 |
Synonym | cobalt ii sulfate hydrate,cobalt sulfate monohydrate,unii-5zqa69klc0,5zqa69klc0,cobalt mesosulfate coh2so5,cobalt sulfate coso4 monohydrate,cobalt ii sulfatehydrate,cobalt sulphate monohydrate,co.so4.h2o,cobalt 2+ hydrate sulfate |
InChI Key | BGORGFZEVHFAQU-UHFFFAOYSA-L |
SMILES | O.[O-]S(=O)(=O)[O-].[Co+2] |
IUPAC Name | cobalt(2+);sulfate;hydrate |
Molecular Weight (g/mol) | 173.004 |
Show More |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects. May cause cancer by inhalation. Harmful if swallowed. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Suspected of causing genetic defects. May damage fertility.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. Wear eye protection/face protection. Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Danger