Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cobalt(II) sulfate hydrate, Reagent, Co 20.8% min

Click to view available options
Quantity:
250g
1kg
60459-08-7
CoH2O5S
173.004
MFCD00149657
BGORGFZEVHFAQU-UHFFFAOYSA-L
cobalt ii sulfate hydrate,cobalt sulfate monohydrate,unii-5zqa69klc0,5zqa69klc0,cobalt mesosulfate coh2so5,cobalt sulfate coso4 monohydrate,cobalt ii sulfatehydrate,cobalt sulphate monohydrate,co.so4.h2o,cobalt 2+ hydrate sulfate
16210984
cobalt(2+);sulfate;hydrate
O.[O-]S(=O)(=O)[O-].[Co+2]
60459-08-7
CoH2O5S
173.004
MFCD00149657
BGORGFZEVHFAQU-UHFFFAOYSA-L
cobalt ii sulfate hydrate,cobalt sulfate monohydrate,unii-5zqa69klc0,5zqa69klc0,cobalt mesosulfate coh2so5,cobalt sulfate coso4 monohydrate,cobalt ii sulfatehydrate,cobalt sulphate monohydrate,co.so4.h2o,cobalt 2+ hydrate sulfate
16210984
cobalt(2+);sulfate;hydrate
O.[O-]S(=O)(=O)[O-].[Co+2]

Ceramics, pigments, glazes, plating baths, catalyst, storage batteries, paint/ink drier

Linear Formula CoSO4·xH2O
UN Number UN3077
Solubility Information Soluble in water; Slightly soluble in methanol and ethanol
Formula Weight 154.99 (Anhydrous)
Physical Form Crystalline
Grade Reagent
Concentration Co 20.8% min.
Color Red to Orange
Quantity 1kg
Chemical Name or Material Cobalt(II) sulfate hydrate

GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.

P201-P202-P261-P264b-P270-P272-P280-P285-P301+P312-P302+P352-P304+P341-P308+P313-P330-P333+P313-P342+P311-P363-P501c

H302-H317-H334-H341-H350-H360-H360F-H500

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 233-334-2

TSCA : Yes

Recommended Storage : Ambient temperatures