Alfa Aesar™ Cobalt iron oxide, may contain small amounts of iron oxide

Manufacturer: Alfa Aesar™ 2314714

Quantity 25g

 View more versions of this product

Catalog No. AA2314714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12052-28-7
    Formula Weight 234.62
    MDL Number MFCD00016018
    Synonym Cobalt ferrite
    Chemical Name or Material Cobalt iron oxide, May contain small amounts of iron oxide

    Specifications

    Formula Weight 234.62
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 12052-28-7
    Synonym Cobalt ferrite
    Quantity 25g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H317
    Toxic if swallowed.
    May cause an allergic skin reaction.
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00016018
    Chemical Name or Material Cobalt iron oxide, May contain small amounts of iron oxide

    GHS H Statement
    H301-H317
    Toxic if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P301+P310-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit